siRNA Details
| Gene Symbol: Sectm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145373.2 | NP_663348.2 | 3 | 454 | ||
| GenBank mRNA | AW985540.1 | - | 3 | 454 | ||
| BC010462.1 | - | 3 | 421 | 3585040 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sectm1a| Gene Symbol: Sectm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145373.2 | NP_663348.2 | 3 | 454 | ||
| GenBank mRNA | AW985540.1 | - | 3 | 454 | ||
| BC010462.1 | - | 3 | 421 | 3585040 | ||