siRNA Details
| Gene Symbol: Tm6sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145375.4 | NP_663350.2 | 1, 2 | 546 | ||
| GenBank mRNA | AK046714.1 | - | 1, 2 | 447 | ||
| AK083132.1 | - | 1, 2 | 549 | |||
| AK135100.1 | - | 1, 2 | 528 | |||
| AK162572.1 | - | 1, 2 | 398 | |||
| AK163621.1 | - | 1, 2 | 471 | |||
| AK172109.1 | - | 1, 2 | 462 | |||
| AK172145.1 | - | 1, 2 | 447 | |||
| BB653227.1 | - | 1, 2 | 548 | |||
| BC023123.1 | - | 1, 2 | 428 | 3995280 | ||

Tm6sf1
Genomic Map