siRNA Details
| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145400.3 | NP_663375.3 | 5 | 565 | ||
| GenBank mRNA | AK028721.1 | - | 5 | 542 | ||
| AK028850.1 | - | 5 | 580 | |||
| AK172893.1 | - | 5 | 525 | |||
| BC006649.1 | - | 6 | 704 | 3498834 | ||
| BC021406.1 | - | 5 | 571 | |||
| BC025043.1 | - | 5 | 534 | |||

Ube4a
Genomic Map