siRNA Details
| Gene Symbol: Cald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145575.3 | NP_663550.1 | 7 | 1368 | ||
| GenBank mRNA | AK148009.1 | - | 5 | 740 | ||
| BC019435.1 | - | 7 | 1327 | 5148205 | ||
| BQ769983.1 | - | 4 | 552 | |||
| CX568419.1 | - | 2 | 64 | |||

Cald1
Genomic Map