siRNA Details
| Gene Symbol: PNMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006029.4 | NP_006020.4 | 1 | 2333 | ||
| GenBank mRNA | AF320308.1 | - | 1 | 1818 | ||
| AL834327.1 | - | 1 | 1066 | |||
| BC039577.1 | - | 1 | 1636 | 4431875 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PNMA1| Gene Symbol: PNMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006029.4 | NP_006020.4 | 1 | 2333 | ||
| GenBank mRNA | AF320308.1 | - | 1 | 1818 | ||
| AL834327.1 | - | 1 | 1066 | |||
| BC039577.1 | - | 1 | 1636 | 4431875 | ||