siRNA Details
| Gene Symbol: Gga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145929.2 | NP_666041.1 | 10 | 1257 | ||
| GenBank mRNA | AK080881.1 | - | 10 | 880 | ||
| AK155195.1 | - | 10 | 897 | |||
| AK159771.1 | - | 10 | 880 | |||
| AK170437.1 | - | 10 | 898 | |||
| AK171534.1 | - | 10 | 914 | |||
| AK185927.1 | - | 3 | 233 | |||
| AK218460.1 | - | 3 | 233 | |||
| BC026802.1 | - | 10 | 1257 | 4223300 | ||
| BC054422.1 | - | 10 | 901 | |||

Gga1
Genomic Map