siRNA Details
| Gene Symbol: AW549877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145930.2 | NP_666042.2 | 6 | 2271 | ||
| GenBank mRNA | AK045350.1 | - | 6 | 2266 | ||
| AK143413.1 | - | 1 | 889 | |||
| AK154940.1 | - | 6 | 2265 | |||
| BC016504.1 | - | 5 | 2149 | 3709876 | ||

AW549877
Genomic Map