siRNA Details
| Gene Symbol: Fanci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145946.2 | NP_666058.2 | 32, 33 | 3607 | ||
| GenBank mRNA | BC027836.1 | - | 30, 31 | 3334 | ||
| BX519311.1 | - | 1, 2 | 91 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fanci| Gene Symbol: Fanci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145946.2 | NP_666058.2 | 32, 33 | 3607 | ||
| GenBank mRNA | BC027836.1 | - | 30, 31 | 3334 | ||
| BX519311.1 | - | 1, 2 | 91 | |||