siRNA Details
| Gene Symbol: HS3ST3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006042.2 | NP_006033.1 | 2 | 2383 | ||
| XM_011524114.2 | XP_011522416.1 | 3 | 2627 | |||
| GenBank mRNA | AF105376.1 | - | 2 | 2368 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS3ST3A1| Gene Symbol: HS3ST3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006042.2 | NP_006033.1 | 2 | 2383 | ||
| XM_011524114.2 | XP_011522416.1 | 3 | 2627 | |||
| GenBank mRNA | AF105376.1 | - | 2 | 2368 | ||