siRNA Details
| Gene Symbol: Kbtbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145958.2 | NP_666070.2 | 4 | 1199 | ||
| GenBank mRNA | AK028396.1 | - | 4 | 1145 | ||
| AK129374.1 | - | 4 | 1109 | |||
| BC022962.1 | - | 1 | 116 | 5368323 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kbtbd2| Gene Symbol: Kbtbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145958.2 | NP_666070.2 | 4 | 1199 | ||
| GenBank mRNA | AK028396.1 | - | 4 | 1145 | ||
| AK129374.1 | - | 4 | 1109 | |||
| BC022962.1 | - | 1 | 116 | 5368323 | ||