siRNA Details
Gene Symbol: CHRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000738.2 | NP_000729.2 | 2 | 562 | ||
XM_011544742.2 | XP_011543044.1 | 2 | 263 | |||
GenBank mRNA | AF385587.1 | - | 1 | 171 | ||
AF498915.1 | - | 1 | 161 | |||
AK312374.1 | - | 2 | 323 | |||
BC007740.2 | - | 2 | 283 | 4128514 | ||
BC022984.1 | - | 2 | 562 | 4931283 | ||
BT007166.1 | - | 1 | 161 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
