siRNA Details
| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147201.2 | NP_671734.1 | 10 | 991 | ||
| GenBank mRNA | AF302138.1 | - | 10 | 990 | ||
| AK046142.1 | - | 11 | 1009 | |||
| AK077440.1 | - | 10 | 1027 | |||
| AK170501.1 | - | 10 | 1007 | |||
| AK188331.1 | - | 2 | 132 | |||
| AK207083.1 | - | 2 | 132 | |||
| AK210846.1 | - | 2 | 133 | |||
| BC004756.1 | - | 10 | 961 | 3154089 | ||
| BC018463.1 | - | 10 | 965 | 3598438 | ||

Nrbp1
Genomic Map