siRNA Details
| Gene Symbol: Foxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148935.2 | NP_683737.2 | 10 | 2047 | ||
| GenBank mRNA | AF323488.1 | - | 10 | 2034 | ||
| AK081735.1 | - | 8 | 1752 | |||
| BC092242.1 | - | 10 | 2036 | 30698162 | ||
| BC130222.1 | - | 10 | 2048 | 40141600 | ||

Foxn4
Genomic Map