siRNA Details
| Gene Symbol: Slc8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148946.2 | NP_683748.1 | 2 | 526 | ||
| GenBank mRNA | AF503502.1 | - | 1 | 391 | ||
| AK035163.1 | - | 2 | 668 | |||
| BC058704.1 | - | 2 | 526 | 6838206 | ||
| BC080277.1 | - | 2 | 478 | 30354551 | ||

Slc8a2
Genomic Map