siRNA Details
Gene Symbol: Pknox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029838.2 | NP_001025009.1 | 7 | 790 | ||
NM_148950.3 | NP_683752.2 | 6 | 685 | |||
GenBank mRNA | AB086817.1 | - | 4 | 511 | ||
AF487460.1 | - | 5 | 591 | |||
AK076428.1 | - | 1 | 1421 | |||
AK082952.1 | - | 7 | 735 | |||
AK084178.1 | - | 7 | 733 | |||
BC050865.1 | - | 6 | 619 | 6332968 |
