siRNA Details
| Gene Symbol: Mmrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153127.3 | NP_694767.3 | 6 | 1361 | ||
| GenBank mRNA | AK077884.1 | - | 6 | 1363 | ||
| AK078470.1 | - | 6 | 1337 | |||
| AK134285.1 | - | 6 | 1400 | |||
| BC145845.1 | - | 6 | 1281 | 40131148 | ||

Mmrn2
Genomic Map