siRNA Details
Gene Symbol: Unc5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153131.3 | NP_694771.1 | 9 | 1585 | ||
GenBank mRNA | AJ487852.1 | - | 9 | 1586 | ||
BC058084.1 | - | 8 | 1438 | 6813463 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
