siRNA Details
Gene Symbol: Irgq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153134.3 | NP_694774.3 | 3 | 2576 | ||
GenBank mRNA | BC056347.1 | - | 3 | 2535 | 5705226 | |
BC080294.1 | - | 3 | 2514 | 6856174 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Irgq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153134.3 | NP_694774.3 | 3 | 2576 | ||
GenBank mRNA | BC056347.1 | - | 3 | 2535 | 5705226 | |
BC080294.1 | - | 3 | 2514 | 6856174 |