siRNA Details
| Gene Symbol: Irgq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153134.3 | NP_694774.3 | 3 | 3592 | ||
| GenBank mRNA | BC056347.1 | - | 3 | 3551 | 5705226 | |
| BC080294.1 | - | 3 | 3530 | 6856174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irgq| Gene Symbol: Irgq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153134.3 | NP_694774.3 | 3 | 3592 | ||
| GenBank mRNA | BC056347.1 | - | 3 | 3551 | 5705226 | |
| BC080294.1 | - | 3 | 3530 | 6856174 | ||