siRNA Details
| Gene Symbol: Unc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153135.3 | NP_694775.1 | 4 | 803 | ||
| GenBank mRNA | AJ487854.1 | - | 4 | 804 | ||
| BC137615.1 | - | 4 | 594 | 8860634 | ||
| BC144783.1 | - | 4 | 594 | 9053331 | ||
| BC144784.1 | - | 4 | 594 | 9053332 | ||
| BC145695.1 | - | 4 | 594 | 40130998 | ||
| FJ824810.1 | - | 4 | 736 | |||

Unc5d
Genomic Map