siRNA Details
| Gene Symbol: Ints9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253731.1 | NP_001240660.1 | 5 | 621 | ||
| NM_153414.4 | NP_700463.2 | 5 | 589 | |||
| GenBank mRNA | AK035769.1 | - | 5 | 333 | ||
| AK038979.1 | - | 5 | 322 | |||
| AK077634.1 | - | 5 | 590 | |||
| AK136992.1 | - | 5 | 316 | |||
| AK150195.1 | - | 5 | 316 | |||
| BC028953.1 | - | 4 | 279 | 5362016 | ||
| BC055700.1 | - | 5 | 288 | 6808816 | ||

Ints9
Genomic Map