siRNA Details
| Gene Symbol: Zfp280c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166648.1 | NP_001160120.1 | 10 | 1003 | ||
| NM_001166649.1 | NP_001160121.1 | 10 | 1024 | |||
| NM_001166650.1 | NP_001160122.1 | 9 | 962 | |||
| NM_153532.3 | NP_705760.2 | 9 | 1334 | |||
| GenBank mRNA | AK129399.1 | - | 10 | 983 | ||
| AK141491.1 | - | 10 | 1004 | |||
| BC028839.1 | - | 9 | 974 | 5150255 | ||
| BC051397.1 | - | 9 | 962 | 5150255 | ||
| BC063775.1 | - | 9 | 1346 | 5375514 | ||

Zfp280c
Genomic Map