siRNA Details
| Gene Symbol: Pygb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153781.1 | NP_722476.1 | 1 | 299 | ||
| GenBank mRNA | AK032769.1 | - | 1 | 312 | ||
| AK035673.1 | - | 1 | 28 | |||
| AK134667.1 | - | 1 | 309 | |||
| AK142716.1 | - | 1 | 308 | |||
| AK147763.1 | - | 1 | 331 | |||
| AK168381.1 | - | 1 | 309 | |||
| AK169631.1 | - | 1 | 313 | |||
| BC031457.1 | - | 1 | 307 | |||
| BC035283.1 | - | 1 | 299 | 4947455 | ||

Pygb
Genomic Map