siRNA Details
| Gene Symbol: Fam210a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153794.4 | NP_722489.1 | 3 | 809 | ||
| GenBank mRNA | AK034980.1 | - | 3 | 691 | ||
| AK044238.1 | - | 3 | 697 | |||
| AK049567.1 | - | 3 | 697 | |||
| AK077093.1 | - | 3 | 810 | |||
| AK142190.1 | - | 3 | 681 | |||
| AK147662.1 | - | 3 | 684 | |||
| AK159319.1 | - | 3 | 660 | |||
| AK162396.1 | - | 3 | 694 | |||
| BC038695.1 | - | 3 | 576 | 1548421 | ||
| BC095936.1 | - | 3 | 659 | 3968496 | ||

Fam210a
Genomic Map