siRNA Details
Gene Symbol: Dio3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172119.2 | NP_742117.2 | 1 | 1685 | ||
GenBank mRNA | AK140797.1 | - | 1 | 1686 | ||
AK146060.1 | - | 1 | 1686 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dio3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172119.2 | NP_742117.2 | 1 | 1685 | ||
GenBank mRNA | AK140797.1 | - | 1 | 1686 | ||
AK146060.1 | - | 1 | 1686 |