siRNA Details
| Gene Symbol: Tbc1d16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172443.3 | NP_766031.2 | 3 | 446 | ||
| GenBank mRNA | AK047555.1 | - | 3 | 447 | ||
| BC057634.1 | - | 3 | 408 | 2866041 | ||
| BQ831250.1 | - | 1 | 29 | |||
| CJ111152.1 | - | 3 | 418 | |||

Tbc1d16
Genomic Map