siRNA Details
| Gene Symbol: Adamts18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172466.2 | NP_766054.2 | 4 | 1111 | ||
| GenBank mRNA | AK035797.1 | - | 4 | 1011 | ||
| AK036137.1 | - | 2 | 277 | |||
| AK053851.1 | - | 3 | 1219 | |||
| AK140876.1 | - | 4 | 887 | |||
| BC094674.1 | - | 4 | 766 | 30548647 | ||
| BC144816.1 | - | 2 | 369 | |||
| BC144817.1 | - | 2 | 368 | |||

Adamts18
Genomic Map