siRNA Details
| Gene Symbol: Zc3hav1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172467.3 | NP_766055.1 | 4 | 929 | ||
| GenBank mRNA | AK045287.1 | - | 4 | 930 | ||
| AK051064.1 | - | 4 | 931 | |||
| AK051832.1 | - | 2 | 317 | |||
| AK084475.1 | - | 4 | 861 | |||
| BC138180.1 | - | 4 | 1021 | 8861199 | ||
| BC138181.1 | - | 4 | 1021 | 8861200 | ||

Zc3hav1l
Genomic Map