siRNA Details
| Gene Symbol: AGFG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006076.4 | NP_006067.3 | 3 | 552 | ||
| XM_005250306.2 | XP_005250363.1 | 3 | 538 | |||
| XM_017012112.1 | XP_016867601.1 | 3 | 493 | |||
| XM_017012113.1 | XP_016867602.1 | 3 | 538 | |||
| XM_017012114.1 | XP_016867603.1 | 3 | 538 | |||
| GenBank mRNA | AF015042.1 | - | 3 | 476 | ||
| AK314954.1 | - | 3 | 538 | |||
| BC009393.2 | - | 3 | 551 | |||
| BC017329.2 | - | 3 | 504 | 4810320 | ||
| BF314987.1 | - | 3 | 551 | |||

AGFG2
Genomic Map