siRNA Details
| Gene Symbol: Tmc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172476.4 | NP_766064.2 | 12 | 1727 | ||
| GenBank mRNA | AK083189.1 | - | 12 | 1730 | ||
| AK132820.1 | - | 12 | 1757 | |||
| AY236499.1 | - | 12 | 1788 | |||
| AY263161.1 | - | 12 | 1756 | |||
| BC105966.1 | - | 6 | 758 | |||

Tmc7
Genomic Map