siRNA Details
| Gene Symbol: Slc38a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172479.3 | NP_766067.2 | 16 | 1600 | ||
| GenBank mRNA | AK054485.1 | - | 16 | 1587 | ||
| AK077667.1 | - | 16 | 1569 | |||
| AK155928.1 | - | 16 | 1553 | |||
| AK158125.1 | - | 15 | 1721 | |||
| BC058856.1 | - | 7 | 858 | |||
| BC152401.1 | - | 16 | 1569 | 40057282 | ||
| BX523289.1 | - | 2 | 247 | |||

Slc38a5
Genomic Map