siRNA Details
| Gene Symbol: Thsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172485.3 | NP_766073.2 | 4 | 1440 | ||
| GenBank mRNA | AK047508.1 | - | 1226 | |||
| AK051714.1 | - | 4 | 1282 | |||
| AK083798.1 | - | 4 | 1268 | |||
| BC063250.1 | - | 4 | 1270 | 6410178 | ||

Thsd7b
Genomic Map