siRNA Details
| Gene Symbol: Prrt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289699.1 | NP_001276628.1 | 2 | 760 | ||
| NM_001289700.1 | NP_001276629.1 | 2 | 681 | |||
| NM_172487.4 | NP_766075.2 | 2 | 734 | |||
| GenBank mRNA | AK031947.1 | - | 2 | 684 | ||
| AK045491.1 | - | 2 | 638 | |||
| AK045678.1 | - | 2 | 722 | |||
| AK136854.1 | - | 2 | 714 | |||
| BC058349.1 | - | 2 | 734 | 5695444 | ||

Prrt3
Genomic Map