siRNA Details
| Gene Symbol: Ncoa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172495.5 | NP_766083.3 | 7 | 952 | ||
| GenBank mRNA | AK128935.1 | - | 6 | 775 | ||
| AK144324.1 | - | 7 | 784 | |||
| BC076623.1 | - | 5 | 558 | |||
| BC141150.1 | - | 6 | 686 | 9055762 | ||
| BC145459.1 | - | 5 | 572 | |||

Ncoa7
Genomic Map