siRNA Details
| Gene Symbol: Phka2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177878.1 | NP_001171349.1 | 9, 10 | 1319 | ||
| NM_001177879.1 | NP_001171350.1 | 10, 11 | 1133 | |||
| NM_172783.3 | NP_766371.1 | 9, 10 | 1319 | |||
| GenBank mRNA | AK052346.1 | - | 9, 10 | 1064 | ||
| AK052992.1 | - | 9, 10 | 1320 | |||
| AK165506.1 | - | 10, 11 | 1134 | |||
| BC050040.1 | - | 10, 11 | 1092 | 6431743 | ||
| BC051167.1 | - | 4, 5 | 559 | |||
| BC058254.1 | - | 9, 10 | 1288 | |||

Phka2
Genomic Map