siRNA Details
| Gene Symbol: Tmem194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113211.1 | NP_001106682.1 | 4, 5 | 577 | ||
| NM_173732.3 | NP_776093.1 | 4, 5 | 577 | |||
| GenBank mRNA | AK090031.1 | - | 4, 5 | 556 | ||
| AK129110.1 | - | 4, 5 | 574 | |||
| AK132974.1 | - | 1046 | ||||
| BC030440.1 | - | 4, 5 | 588 | 5373527 | ||

Tmem194
Genomic Map