siRNA Details
| Gene Symbol: CHRM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000741.3 | NP_000732.2 | 1 | 929 | ||
| GenBank mRNA | AF385590.1 | - | 1 | 878 | ||
| AF498918.1 | - | 1 | 878 | |||
| BC095546.1 | - | 1 | 879 | |||
| BC119775.1 | - | 1 | 877 | |||
| BC119817.1 | - | 1 | 877 | |||
| BC137535.1 | - | 1 | 929 | 9021535 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007699, BC137677, S74916, BC120504.
Set Membership: |
|

CHRM4
Genomic Map