siRNA Details
| Gene Symbol: Fmr1nb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174993.2 | NP_778158.2 | 3 | 523 | ||
| GenBank mRNA | AY214130.1 | - | 3 | 428 | ||
| BC145139.1 | - | 3 | 504 | 9053689 | ||
| BC145140.1 | - | 3 | 504 | 9053690 | ||
| BC171947.1 | - | 3 | 504 | 9054233 | ||
| BC171948.1 | - | 3 | 504 | 9054234 | ||

Fmr1nb
Genomic Map