siRNA Details
Gene Symbol: Srl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175347.4 | NP_780556.2 | 2 | 298 | ||
GenBank mRNA | AB113383.1 | - | 2 | 298 | ||
BC145687.1 | - | 2 | 320 | 40130990 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Srl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175347.4 | NP_780556.2 | 2 | 298 | ||
GenBank mRNA | AB113383.1 | - | 2 | 298 | ||
BC145687.1 | - | 2 | 320 | 40130990 |