siRNA Details
| Gene Symbol: Catsperd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175350.3 | NP_780559.2 | 19 | 1684 | ||
| GenBank mRNA | AK077082.1 | - | 10 | 745 | ||
| BC115864.1 | - | 8 | 637 | |||
| BC116633.1 | - | 9 | 751 | 40056655 | ||
| HQ441159.1 | - | 19 | 1676 | |||
| HQ441160.1 | - | 16 | 1572 | |||

Catsperd
Genomic Map