siRNA Details
| Gene Symbol: Cdca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110162.1 | NP_001103632.1 | 6 | 1003 | ||
| NM_175384.4 | NP_780593.3 | 6 | 706 | |||
| GenBank mRNA | AK031083.1 | - | 6 | 769 | ||
| AK031313.1 | - | 2 | 148 | |||
| AK160223.1 | - | 6 | 765 | |||
| AK166952.1 | - | 6 | 772 | |||
| BC037136.1 | - | 6 | 803 | |||
| BC067009.1 | - | 6 | 734 | |||
| BC075682.1 | - | 6 | 706 | |||
| BC116298.1 | - | 2 | 148 | 40095010 | ||
| BC116299.1 | - | 2 | 148 | 40095011 | ||

Cdca2
Genomic Map