siRNA Details
| Gene Symbol: Cdca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110162.1 | NP_001103632.1 | 8, 9 | 1415 | ||
| NM_175384.4 | NP_780593.3 | 8, 9 | 1118 | |||
| GenBank mRNA | AK031313.1 | - | 4, 5 | 560 | ||
| AK160223.1 | - | 8, 9 | 1177 | |||
| AK166952.1 | - | 8, 9 | 1185 | |||
| AK208630.1 | - | 1, 2 | 75 | |||
| AK208800.1 | - | 1, 2 | 75 | |||
| BC037136.1 | - | 8, 9 | 1215 | |||
| BC067009.1 | - | 8, 9 | 1146 | |||
| BC075682.1 | - | 8, 9 | 1118 | |||
| BC116298.1 | - | 4, 5 | 560 | 40095010 | ||
| BC116299.1 | - | 4, 5 | 560 | 40095011 | ||

Cdca2
Genomic Map