siRNA Details
| Gene Symbol: Fam73b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242407.1 | NP_001229336.1 | 4 | 568 | ||
| NM_175392.3 | NP_780601.1 | 4 | 736 | |||
| GenBank mRNA | AK077618.1 | - | 4 | 529 | ||
| AK131152.1 | - | 1 | 1422 | |||
| AK140122.1 | - | 4 | 554 | |||
| AK154200.1 | - | 4 | 557 | |||
| AK165996.1 | - | 4 | 559 | |||
| AK210993.1 | - | 2 | 79 | |||
| AK218176.1 | - | 2 | 99 | |||
| BC051404.1 | - | 4 | 736 | 6395030 | ||

Fam73b
Genomic Map