siRNA Details
| Gene Symbol: A930024E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042918.1 | - | 1 | 697 | ||
| AK079939.1 | - | 1 | 85 | |||
| AK080729.1 | - | 1 | 604 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930024E05Rik| Gene Symbol: A930024E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042918.1 | - | 1 | 697 | ||
| AK079939.1 | - | 1 | 85 | |||
| AK080729.1 | - | 1 | 604 | |||