siRNA Details
| Gene Symbol: C1qtnf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135172.1 | NP_001128644.1 | 3 | 1484 | ||
| NM_175425.4 | NP_780634.2 | 3 | 1127 | |||
| GenBank mRNA | AK017441.1 | - | 3 | 1476 | ||
| AK078818.1 | - | 3 | 1290 | |||
| BC038308.1 | - | 3 | 1127 | |||
| BC090967.1 | - | 3 | 1465 | 30652983 | ||
| DQ002400.1 | - | 3 | 1136 | |||

C1qtnf7
Genomic Map