siRNA Details
| Gene Symbol: Fam163b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175427.4 | NP_780636.1 | 3 | 1091 | ||
| GenBank mRNA | AK083284.1 | - | 3 | 1089 | ||
| AK162827.1 | - | 3 | 1092 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam163b| Gene Symbol: Fam163b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175427.4 | NP_780636.1 | 3 | 1091 | ||
| GenBank mRNA | AK083284.1 | - | 3 | 1089 | ||
| AK162827.1 | - | 3 | 1092 | |||