siRNA Details
| Gene Symbol: Zbtb21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081684.1 | NP_001075153.1 | 2 | 3777 | ||
| NM_001081685.1 | NP_001075154.1 | 4 | 3320 | |||
| NM_175428.3 | NP_780637.2 | 3 | 3920 | |||
| GenBank mRNA | AK030829.1 | - | 2 | 1853 | ||
| AK034057.1 | - | 3 | 3922 | |||
| AK129313.1 | - | 2 | 3754 | |||

Zbtb21
Genomic Map