siRNA Details
| Gene Symbol: Fam228b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175431.4 | NP_780640.2 | 5, 6 | 844 | ||
| GenBank mRNA | AK044133.1 | - | 5, 6 | 845 | ||
| AK050447.1 | - | 5, 6 | 810 | |||
| BC100429.1 | - | 5, 6 | 623 | 30917787 | ||
| BC145648.1 | - | 5, 6 | 575 | 40130951 | ||

Fam228b
Genomic Map