siRNA Details
Gene Symbol: Zfp710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145999.1 | NP_001139471.1 | 5 | 4344 | ||
NM_001146000.1 | NP_001139472.1 | 6 | 4442 | |||
NM_175433.5 | NP_780642.3 | 5 | 4427 | |||
GenBank mRNA | AK036711.1 | - | 4 | 3004 | ||
AK154953.1 | - | 5 | 4345 | |||
BC058640.1 | - | 5 | 4238 | 6835601 | ||
BC060641.1 | - | 5 | 4321 | 6849964 | ||
CN459717.1 | - | 1 | 142 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
