siRNA Details
| Gene Symbol: Osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176840.3 | NP_789810.2 | 4 | 1206 | ||
| GenBank mRNA | BC035278.1 | - | 4 | 1206 | 5043236 | |
| BC099400.1 | - | 4 | 1121 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbpl11| Gene Symbol: Osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176840.3 | NP_789810.2 | 4 | 1206 | ||
| GenBank mRNA | BC035278.1 | - | 4 | 1206 | 5043236 | |
| BC099400.1 | - | 4 | 1121 | |||